6-Bromo-2-(dimethoxymethyl)furo[3,2-b]pyridine

Supplier: Matrix Scientific

048426-250MG
101883-934EA 393.3 USD
101883-934
6-Bromo-2-(dimethoxymethyl)furo[3,2-b]pyridine
6-Bromo-2-(dimethoxymethyl)furo[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₀H₁₀BrNO₃
MDL Number: MFCD12922781
CAS Number: 1186311-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR