2-((tert-Butyldimethylsilyloxy)methyl)-6-iodofuro[3,2-b]pyridine

Supplier: Matrix Scientific

048424-250MG
101883-930EA 393.3 USD
101883-930
2-((tert-Butyldimethylsilyloxy)methyl)-6-iodofuro[3,2-b]pyridine
2-((tert-Butyldimethylsilyloxy)methyl)-6-iodofuro[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₄H₂₀INO₂Si
MDL Number: MFCD12401659
CAS Number: 1171920-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR