Furo[3,2-b]pyridin-6-ol

Supplier: Matrix Scientific

048419-250MG
101883-920EA 368 USD
101883-920
Furo[3,2-b]pyridin-6-ol
Furo[3,2-b]pyridin-6-ol

Hazard: Irritant

Formula: C₇H₅NO₂
MDL Number: MFCD12401654
CAS Number: 1171920-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR