4-(Dimethoxymethyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

048418-250MG
101883-918EA 526.15 USD
101883-918
4-(Dimethoxymethyl)-1H-pyrrolo[2,3-b]pyridine
4-(Dimethoxymethyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MDL Number: MFCD12922780
CAS Number: 1186311-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR