5-Fluoro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Supplier: Matrix Scientific
048417-250MG
101883-916EA
492.31
USD
101883-916
5-Fluoro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
5-Fluoro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Hazard: Irritant
Formula:
C₈H₅FN₂O |
MDL Number:
MFCD12922779 CAS Number: 1171920-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...