4,5-Dichloro-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

048414-250MG
101883-910EA 526.15 USD
101883-910
4,5-Dichloro-1H-pyrrolo[2,3-b]pyridine
4,5-Dichloro-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₄Cl₂N₂
MDL Number: MFCD12401653
CAS Number: 1142192-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR