6-Bromo-1,8-naphthyridin-2-ol

Supplier: Matrix Scientific

048411-250MG
101883-904EA 393.3 USD
101883-904
6-Bromo-1,8-naphthyridin-2-ol
6-Bromo-1,8-naphthyridin-2-ol

Hazard: Irritant

Formula: C₈H₅BrN₂O
MDL Number: MFCD07781151
CAS Number: 72754-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR