5-Bromo-3-((3-((tert-butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)-2-chloropyridine

Supplier: Matrix Scientific

048409-250MG
101883-900EA 393.3 USD
101883-900
5-Bromo-3-((3-((tert-butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)-2-chloropyridine
5-Bromo-3-((3-((tert-butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)-2-chloropyridine

Hazard: Irritant

Formula: C₁₇H₂₈BrClN₂OSi
MDL Number: MFCD12922773
CAS Number: 1186311-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR