3-((3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)-5-iodopyridine

Supplier: Matrix Scientific

048407-250MG
101883-896EA 393.3 USD
101883-896
3-((3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)-5-iodopyridine
3-((3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)-5-iodopyridine

Hazard: Irritant

Formula: C₁₇H₂₉IN₂OSi
MDL Number: MFCD12922771
CAS Number: 1186311-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR