N-(3-Allylpyridin-4-yl)pivalamide

Supplier: Matrix Scientific

048395-250MG
101883-872EA 526.15 USD
101883-872
N-(3-Allylpyridin-4-yl)pivalamide
N-(3-Allylpyridin-4-yl)pivalamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
MDL Number: MFCD12922759
CAS Number: 1186311-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR