2-Chloro-N-(2-hydroxypyridin-3-yl)acetamide

Supplier: Matrix Scientific

048391-250MG
101883-864EA 393.3 USD
101883-864
2-Chloro-N-(2-hydroxypyridin-3-yl)acetamide
2-Chloro-N-(2-hydroxypyridin-3-yl)acetamide

Hazard: Irritant

Formula: C₇H₇ClN₂O₂
MDL Number: MFCD12922757
CAS Number: 112777-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR