N-(2-Hydroxypyridin-3-yl)acetamide

Supplier: Matrix Scientific

048388-250MG
101883-858EA 393.3 USD
101883-858
N-(2-Hydroxypyridin-3-yl)acetamide
N-(2-Hydroxypyridin-3-yl)acetamide

Hazard: Irritant

Formula: C₇H₈N₂O₂
MDL Number: MFCD12922754
CAS Number: 76349-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR