3-(Dimethoxymethyl)-5-iodopyridine

Supplier: Matrix Scientific

048387-250MG
101883-856EA 393.3 USD
101883-856
3-(Dimethoxymethyl)-5-iodopyridine
3-(Dimethoxymethyl)-5-iodopyridine

Hazard: Irritant

Formula: C₈H₁₀INO₂
MDL Number: MFCD12922753
CAS Number: 1186311-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR