3-Bromo-5-(dimethoxymethyl)pyridine

Supplier: Matrix Scientific

048386-250MG
101883-854EA 526.15 USD
101883-854
3-Bromo-5-(dimethoxymethyl)pyridine
3-Bromo-5-(dimethoxymethyl)pyridine

Hazard: Irritant

Formula: C₈H₁₀BrNO₂
MDL Number: MFCD10699139
CAS Number: 163163-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR