5-Bromo-2-morpholinopyridin-3-amine

Supplier: Matrix Scientific

048385-250MG
101883-852EA 393.3 USD
101883-852
5-Bromo-2-morpholinopyridin-3-amine
5-Bromo-2-morpholinopyridin-3-amine

Hazard: Irritant

Formula: C₉H₁₂BrN₃O
MDL Number: MFCD08692325
CAS Number: 1186310-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR