N-(2-Hydroxy-5-iodopyridin-3-yl)acetamide

Supplier: Matrix Scientific

048382-250MG
101883-846EA 526.15 USD
101883-846
N-(2-Hydroxy-5-iodopyridin-3-yl)acetamide
N-(2-Hydroxy-5-iodopyridin-3-yl)acetamide

Hazard: Irritant

Formula: C₇H₇IN₂O₂
MDL Number: MFCD12922750
CAS Number: 1186310-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR