5,6-Dimethoxynicotinic acid

Supplier: Matrix Scientific

048380-250MG
101883-842EA 391 USD
101883-842
5,6-Dimethoxynicotinic acid
5,6-Dimethoxynicotinic acid

Hazard: Irritant

Formula: C₈H₉NO₄
MDL Number: MFCD12401642
CAS Number: 76470-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR