Methyl-3-(5,6-dimethoxypyridin-3-yl)acrylate

Supplier: Matrix Scientific

048378-250MG
101883-838EA 393.3 USD
101883-838
Methyl-3-(5,6-dimethoxypyridin-3-yl)acrylate
Methyl-3-(5,6-dimethoxypyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD12401640
CAS Number: 1171919-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR