5-Bromo-N,3-dimethoxy-N-methylpicolinamide
Supplier: Matrix Scientific
048377-250MG
101883-836EA
526.15
USD
101883-836
5-Bromo-N,3-dimethoxy-N-methylpicolinamide
5-Bromo-N,3-dimethoxy-N-methylpicolinamide
Hazard: Irritant
Formula:
C₉H₁₁BrN₂O₃ |
MDL Number:
MFCD12401639 CAS Number: 1171919-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...