3-(5,6-Dimethoxypyridin-3-yl)prop-2-yn-1-ol

Supplier: Matrix Scientific

048376-250MG
101883-834EA 393.3 USD
101883-834
3-(5,6-Dimethoxypyridin-3-yl)prop-2-yn-1-ol
3-(5,6-Dimethoxypyridin-3-yl)prop-2-yn-1-ol

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD12401638
CAS Number: 1171919-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR