2,3-Dimethoxy-5-((trimethylsilyl)ethynyl)pyridine

Supplier: Matrix Scientific

048375-250MG
101883-832EA 393.3 USD
101883-832
2,3-Dimethoxy-5-((trimethylsilyl)ethynyl)pyridine
2,3-Dimethoxy-5-((trimethylsilyl)ethynyl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₇NO₂Si
MDL Number: MFCD12401637
CAS Number: 1171919-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR