4-Amino-2-iodopyridin-3-ol

Supplier: Matrix Scientific

048371-250MG
101883-824EA 393.3 USD
101883-824
4-Amino-2-iodopyridin-3-ol
4-Amino-2-iodopyridin-3-ol

Hazard: Irritant

Formula: C₅H₅IN₂O
MDL Number: MFCD12025486
CAS Number: 1171919-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR