N-(2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)pivalamide

Supplier: Matrix Scientific

048370-250MG
101883-822EA 526.15 USD
101883-822
N-(2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)pivalamide
N-(2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₆H₂₄BBrN₂O₃
MDL Number: MFCD12401634
CAS Number: 1309980-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR