6-Bromo-2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Supplier: Matrix Scientific

048367-250MG
101883-816EA 393.3 USD
101883-816
6-Bromo-2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
6-Bromo-2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Hazard: Irritant

Formula: C₁₁H₁₄BBrClNO₂
MDL Number: MFCD12401631
CAS Number: 1247726-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR