3-Iodopyridin-4-ol

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-3-iodopyridine

048364-500MG
101883-810EA 356.5 USD
101883-810
3-Iodopyridin-4-ol
3-Iodopyridin-4-ol

Hazard: Irritant

Formula: C₅H₄INO
MW: 221 g/mol
MDL Number: MFCD01646059
CAS Number: 89282-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR