tert-Butyl-3-((3-iodopyridin-2-yloxy)methyl)pyrrolidine-1-carboxylate

Supplier: Matrix Scientific

048363-250MG
101883-808EA 393.3 USD
101883-808
tert-Butyl-3-((3-iodopyridin-2-yloxy)methyl)pyrrolidine-1-carboxylate
tert-Butyl-3-((3-iodopyridin-2-yloxy)methyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₁IN₂O₃
MDL Number: MFCD12922746
CAS Number: 1186310-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR