N-(3-Hydroxy-2-iodopyridin-4-yl)acetamide

Supplier: Matrix Scientific

048362-250MG
101883-806EA 393.3 USD
101883-806
N-(3-Hydroxy-2-iodopyridin-4-yl)acetamide
N-(3-Hydroxy-2-iodopyridin-4-yl)acetamide

Hazard: Irritant

Formula: C₇H₇IN₂O₂
MDL Number: MFCD12922745
CAS Number: 1186310-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR