2-((Trimethylsilyl)ethynyl)pyridin-3-amine

Supplier: Matrix Scientific

048358-250MG
101883-798EA 393.3 USD
101883-798
2-((Trimethylsilyl)ethynyl)pyridin-3-amine
2-((Trimethylsilyl)ethynyl)pyridin-3-amine

Hazard: Irritant

Formula: C₁₀H₁₄N₂Si
MDL Number: MFCD12922741
CAS Number: 947330-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR