3-(2-Chloro-3-(dimethoxymethyl)pyridin-4-yl)prop-2-yn-1-ol
Supplier: Matrix Scientific
048353-250MG
101883-788EA
393.3
USD
101883-788
3-(2-Chloro-3-(dimethoxymethyl)pyridin-4-yl)prop-2-yn-1-ol
3-(2-Chloro-3-(dimethoxymethyl)pyridin-4-yl)prop-2-yn-1-ol
Hazard: Irritant
Formula:
C₁₁H₁₂ClNO₃ |
MDL Number:
MFCD12922736 CAS Number: 1186310-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...