2-Chloro-3-(dimethoxymethyl)isonicotinonitrile

Supplier: Matrix Scientific

048351-250MG
101883-784EA 526.15 USD
101883-784
2-Chloro-3-(dimethoxymethyl)isonicotinonitrile
2-Chloro-3-(dimethoxymethyl)isonicotinonitrile

Hazard: Irritant

Formula: C₉H₉ClN₂O₂
MDL Number: MFCD12922734
CAS Number: 1186310-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR