(E)-4-Chloronicotinaldehyde oxime

Supplier: Matrix Scientific

048346-250MG
101883-774EA 526.15 USD
101883-774
(E)-4-Chloronicotinaldehyde oxime
(E)-4-Chloronicotinaldehyde oxime

Hazard: Irritant

Formula: C₆H₅ClN₂O
MDL Number: MFCD12922730
CAS Number: 471909-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR