tert-Butyl (5-pivalamidopyridin-3-yl)methylcarbamate

Supplier: Matrix Scientific

048345-250MG
101883-772EA 393.3 USD
101883-772
tert-Butyl (5-pivalamidopyridin-3-yl)methylcarbamate
tert-Butyl (5-pivalamidopyridin-3-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₆H₂₅N₃O₃
MDL Number: MFCD12922729
CAS Number: 1171920-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR