tert-Butyl-3-((3-iodopyridin-4-yloxy)methyl)pyrrolidine-1-carboxylate

Supplier: Matrix Scientific

048344-250MG
101883-770EA 526.15 USD
101883-770
tert-Butyl-3-((3-iodopyridin-4-yloxy)methyl)pyrrolidine-1-carboxylate
tert-Butyl-3-((3-iodopyridin-4-yloxy)methyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₁IN₂O₃
MW: 404.25 g/mol
MDL Number: MFCD12922728
CAS Number: 1171920-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR