N-(2-Bromo-5-cyanopyridin-3-yl)pivalamide

Supplier: Matrix Scientific

048342-250MG
101883-766EA 393.3 USD
101883-766
N-(2-Bromo-5-cyanopyridin-3-yl)pivalamide
N-(2-Bromo-5-cyanopyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₂BrN₃O
MDL Number: MFCD12401650
CAS Number: 1171920-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR