3-Iodo-4-methoxypyridine

Supplier: Matrix Scientific

048337-250MG
101883-756EA 107.69 USD
101883-756
3-Iodo-4-methoxypyridine
3-Iodo-4-methoxypyridine

Hazard: Irritant

Formula: C₆H₆INO
MW: 235.0200 g/mol
MDL Number: MFCD01416617
CAS Number: 89640-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR