N-(3-Hydroxy-2-methoxypyridin-4-yl)pivalamide

Supplier: Matrix Scientific

048336-250MG
101883-754EA 526.15 USD
101883-754
N-(3-Hydroxy-2-methoxypyridin-4-yl)pivalamide
N-(3-Hydroxy-2-methoxypyridin-4-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃
MDL Number: MFCD12401648
CAS Number: 1171919-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR