5-Fluoro-3-iodopyridin-2-ol

Supplier: Matrix Scientific

048331-250MG
101883-744EA 364.62 USD
101883-744
5-Fluoro-3-iodopyridin-2-ol
5-Fluoro-3-iodopyridin-2-ol

Hazard: Irritant

Formula: C₅H₃FINO
MW: 238.99 g/mol
MDL Number: MFCD12922725
CAS Number: 1186311-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR