3,4-Dihydro-1H-benzo[e][1,4]diazepine-2,5-dione

Supplier: Matrix Scientific

048324-1G
101883-726EA 229.23 USD
101883-726
3,4-Dihydro-1H-benzo[e][1,4]diazepine-2,5-dione
3,4-Dihydro-1H-benzo[e][1,4]diazepine-2,5-dione

Hazard: Irritant

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
MDL Number: MFCD00111349
CAS Number: 5118-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR