3-Thiophen-2-ylpiperazine-1-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-(thien-2-yl)-piperazine-1-carboxylate

048321-250MG
101883-720EA 264.5 USD
101883-720
3-Thiophen-2-ylpiperazine-1-carboxylic acid ethyl ester ≥95%
3-Thiophen-2-ylpiperazine-1-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂S
MW: 240.33 g/mol
MDL Number: MFCD06245431
CAS Number: 85803-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR