2-(Trifluoromethyl)quinoxaline ≥97%

Supplier: Matrix Scientific

048318-1G
101883-712EA 44.85 USD
101883-712
2-(Trifluoromethyl)quinoxaline ≥97%
2-(Trifluoromethyl)quinoxaline

Hazard: Irritant

Formula: C₉H₅F₃N₂
MDL Number: MFCD06656411
CAS Number: 148853-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR