3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-Boc-3-benzylpiperidine-3-carboxylate

048311-1G
101883-696EA 111.55 USD
101883-696
3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester ≥95%
3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester

Hazard: Irritant

Formula: C₂₀H₂₉NO₄
MW: 347.46 g/mol
MDL Number: MFCD11500859
CAS Number: 170842-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR