1-Boc-DL-nipecotic acid ethyl ester ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 1-Boc-3-piperidinecarboxylate
, Ethyl 1-Boc-DL-nipecotate
048310-1G
101883-694EA
15.38
USD
101883-694
1-Boc-DL-nipecotic acid ethyl ester ≥97%
1-Boc-DL-nipecotic acid ethyl ester
Hazard: Irritant
Formula:
C₁₃H₂₃NO₄ MW: 257.33 g/mol Melting Pt: 31 °C |
MDL Number:
MFCD04116274 CAS Number: 130250-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...