2-Amino-4-(aminosulfonyl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

048303-500MG
101883-680EA 869.23 USD
101883-680
2-Amino-4-(aminosulfonyl)benzenecarboxylic acid ≥95%
2-Amino-4-(aminosulfonyl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₇H₈N₂O₄S
MDL Number: MFCD12025949
CAS Number: 25096-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR