(R)-(+)-Chlocyphos ≥99%
Supplier: Matrix Scientific
Synonyms:
(4R)-(+)-4-(2-Chlorophenyl)-5,5-dimethyl-2-hydroxy-1,3,2-dioxaphosphinane 2-oxide
, (4R)-4-(2-Chlorophenyl)-2-hydroxy-5,5-dimethyl-1,3,2-dioxaphosphinane 2-oxide
048292-500MG
048292-1G
101883-636EA
221.54
USD
101883-636
101883-634
(R)-(+)-Chlocyphos ≥99%
(R)-(+)-Chlocyphos
Hazard: Irritant
Formula:
C₁₁H₁₄ClO₄P MW: 276.66 g/mol |
MDL Number:
MFCD00069409 CAS Number: 98674-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...