2-Amino-4-(benzylsulfanyl)-2-methylbutanoic acid ≥95%

Supplier: Matrix Scientific

048281-500MG 048281-1G
101883-590EA 416.3 USD
101883-590 101883-592
2-Amino-4-(benzylsulfanyl)-2-methylbutanoic acid ≥95%
2-Amino-4-(benzylsulfanyl)-2-methylbutanoic acid

Hazard: Irritant

Formula: C₁₂H₁₇NO₂S
MDL Number: MFCD11553078
CAS Number: 19361-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR