5-Fluoro-2-methyl-1H-indene-3-acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (5-Fluoro-2-methyl-1H-inden-3-yl)acetic acid

048272-1G
101883-562EA 1163.08 USD
101883-562
5-Fluoro-2-methyl-1H-indene-3-acetic acid ≥95%
5-Fluoro-2-methyl-1H-indene-3-acetic acid

Hazard: Irritant

Formula: C₁₂H₁₁FO₂
MW: 206.216 g/mol
MDL Number: MFCD01076194
CAS Number: 32004-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR