2,4-Dioxo-1,4-dihydro-2H-3,1-benzoxazine-8-carboxylic acid ≥95%

Supplier: Matrix Scientific

048266-1G 048266-500MG
101883-540EA 172.5 USD
101883-540 101883-542
2,4-Dioxo-1,4-dihydro-2H-3,1-benzoxazine-8-carboxylic acid ≥95%
2,4-Dioxo-1,4-dihydro-2H-3,1-benzoxazine-8-carboxylic acid

Hazard: Irritant

Formula: C₉H₅NO₅
MW: 207.14 g/mol
MDL Number: MFCD09865033
CAS Number: 167902-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR