3-(Benzoylsulfanyl)-2-methylpropanoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Benzoylthio)-2-methylpropanoic acid
048262-1G
048262-500MG
048262-5G
101883-522EA
806.15
USD
101883-522
101883-520
101883-518
3-(Benzoylsulfanyl)-2-methylpropanoic acid ≥95%
3-(Benzoylsulfanyl)-2-methylpropanoic acid
Hazard: Irritant
Formula:
C₁₁H₁₂O₃S MW: 224.28 g/mol |
MDL Number:
MFCD09864827 CAS Number: 74431-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...