1-(2-Methoxyphenyl)-3-methylpiperazine ≥95%

Supplier: Matrix Scientific

048256-500MG
101883-492EA 208.15 USD
101883-492
1-(2-Methoxyphenyl)-3-methylpiperazine ≥95%
1-(2-Methoxyphenyl)-3-methylpiperazine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD06804057
CAS Number: 152943-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR