2-Cyclobutyl-2-hydroxy-2-phenylacetic acid ≥95%

Supplier: Matrix Scientific

048253-500MG
101883-484EA 529 USD
101883-484
2-Cyclobutyl-2-hydroxy-2-phenylacetic acid ≥95%
2-Cyclobutyl-2-hydroxy-2-phenylacetic acid

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD00037149
CAS Number: 1460-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR